missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm

Accessories for filter holders for sample preparation.

Brand:  Merck XX4204708

 View more versions of this product

Product Code. 10634552

  • £472.00 / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700

SS Filter, Depends on filter type used.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Glass-filled Polypropylene
Back-pressure Screen
25 mm
In-Line Stainless Steel Filter Holder (47 mm)
Non-sterile
1 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm > Glass-Filled Polypropylene Vent/Relief Valve

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.