missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Accessories for filter holders for sample preparation.
£55.00 - £745.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
For Use With (Equipment) | In-Line Stainless Steel Filter Holder (47 mm) |
Sterility | Non-sterile |
Product Code | Brand | Type | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | |||||
10729591
|
Merck
XX4404703 |
Back-pressure Screen | Stainless Steel |
£117.00
Each |
Estimated Shipment: 01-05-2025
Log in to see stock. |
|||||
10673012
|
Merck
XX4304701 |
O-Ring | Silicone |
£181.00
Pack of 6 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10419532
|
Merck
XX4504704 |
Filter Support Screen | Stainless Steel |
£458.00
Each |
Estimated Shipment: 01-05-2025
Log in to see stock. |
|||||
10029810
|
Merck
XX4404702 |
HP Support Screen | PTFE-coated Stainless Steel |
£745.00
Each |
Estimated Shipment: 01-05-2025
Log in to see stock. |
|||||
10634552
|
Merck
XX4204708 |
Back-pressure Screen | Glass-filled Polypropylene |
£472.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
|||||
10460103
|
Merck
XX4204709 |
Stirred Cell Support Screen | Stainless Steel |
£85.00
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10741263
|
Merck
5614 |
Underdrain Screen | Stainless Steel (316) |
£55.00
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
Description
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700SS Filter, † Depends on filter type used.
Specifications
Not Autoclavable | |
In-Line Stainless Steel Filter Holder (47 mm) | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm