Learn More
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Accessories for filter holders for sample preparation.
Brand:  Merck XX4504704
 View more versions of this product
Product Code. 10419532
  • £458.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700
SS Filter, Depends on filter type used.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Stainless Steel
Filter Support Screen
In-Line Stainless Steel Filter Holder (47 mm)
Non-sterile
47 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm > Filter Support Screen