Learn More
BD Emerald™ Three-Part Syringe
Combines optimum performance and reduced environmental impact. BD™ Emerald™ Three-Part Syringe is a general purpose syringe that contains up to 30% less material than other syringes to minimize waste. Syringes feature a center-mounted Luer connection and no needle.
£5.65 - £96.77
Specifications
Graduated Yes
View More Specs
Products 4
Product Code Brand Volume (Metric) Graduations Price Quantity & Availability  
Product Code Brand Volume (Metric) Graduations Price Quantity & Availability  
15285083
View Documents
BD
307727
2 mL 0.1 mL
£96.77
Pack of 100
15275083
View Documents
BD
302986
3 mL 0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15295083
View Documents
BD
307731
5 mL 0.2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15205093
View Documents
BD
307736
10 mL 0.2 mL
£72.24
Pack of 100
Description
Description
  • Combines optimum performance and reduced environmental impact
  • Manufactured in a production plant powered by renewable energy
  • Crystal clear plastic allows for easy viewing of the contents
  • Green-colored piston crown and engraved shell pressure for precise and safe dosage
  • Robust piston stop prevents the rod from accidentally being knocked out
  • Rod designed for stable and comfortable grip during use
  • Thumb grip with the structure for secure grip during administration
Specifications
Specifications
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BD Emerald™ Three-Part Syringe