Learn More
Cole-Parmer™ Endecotts™ Stainless Steel Consistometer
Use this consistometer to determine the consistency of viscous materials by measuring the distance that the material flows under its own weight in a given time interval. Cole-Parmer™ Endecotts™ Stainless Steel Consistometer checks against consistency, viscosity and flow rate standards.
Brand: Cole-Parmer™ 59950-00
Description
Use this consistometer to determine the consistency of viscous materials by measuring the distance that the material flows under its own weight in a given time interval. Cole-Parmer™ Endecotts™ Stainless Steel Consistometer checks against consistency, viscosity and flow rate standards.
- Verify the consistency, viscosity and flow rates of fluids
- Meets military specification R-81294B for paint manufacturing
- Ideal for predetermining product formulas to standardize production lots
- Consistometer requires only 75mL of sample for measurement
- Engraved graduations in 0.5cm divisions ensure accurate results
- Spring-loaded gate prevents premature sample flow
- Stainless steel construction prevents corrosion
Specifications
Consistometer | |
14 cm | |
8.9 cm |
Stainless Steel | |
35.6 cm | |
75 mL |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.