missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimcare Industrie™ Premier Hand Cleanser

Ideal for refilling dispensers in busy working environments. Kimcare Industries™ PREMIER Hand Cleanser is a cost-effective and professional solution to maintaining hygiene and cleanliness.

Brand:  Kimberly-Clark™ 9522

100.98 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12681346

  • £169.00 / Pack of 2
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Green, lightly scented liquid gel hand cleanser
  • Advanced formula removes heavy oil, grease, tar and most inks
  • Contains Aloe Vera and other natural moisturisers to help nourish the skin
  • Cost-effective solution
  • For busy working environments
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hand Cleanser
ISO 14001, ISO 9001
Ideal for removing heavy oil, grease, tar and most inks.
10.6 in.
2 Pk.
3.5 L
Green
24.4 x 17.23 x 13.2 cm
Kimberly-Clark Professional Hand Cleanser Dispenser
26.8 cm
118.3 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Kimcare Industrie™ Premier Hand Cleanser > Capacity: 3.5 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.