Learn More
Tocris Bioscience™ Entecavir
Potent and selective hepatitis B virus inhibitor
Brand:  Tocris Bioscience™ 6234/50
 View more versions of this product
Product Code. 18768209
  • £189.00 / 50mg
Explore more special offers
Description
Description
Entecavir is a potent and selective hepatitis B virus reverse transcriptase inhibitor (EC50 = 3 nM); guanine analog. Inhibits hepatitis B virus replication in HepG2.2.15 cells. Displays selectively over HCMV, HSV-1, VZV, HIV and influenza (EC50 values are 15, 32, 30-60, >10 and >80 μM respectively). Orally bioavailable.
Specifications
Specifications
Inhibitor
C12H15N5O3
BMS 200475, SQ 34676
RNA/DNA Polymerase Inhibitors
142217-69-4
277.28
Store at -20°C