missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Entecavir

Potent and selective hepatitis B virus inhibitor

£70.00 - £189.00

Specifications

Product Type Inhibitor
CAS 142217-69-4
Molecular Formula C12H15N5O3
Formula Weight 277.28
Synonym BMS 200475, SQ 34676
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18770478
View Documents
Tocris Bioscience™
6234/10
10 mg
£70.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18768209
View Documents
Tocris Bioscience™
6234/50
50 mg
£189.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and selective hepatitis B virus reverse transcriptase inhibitor (EC50 = 3 nM); guanine analog. Inhibits hepatitis B virus replication in HepG2.2.15 cells. Displays selectively over HCMV, HSV-1, VZV, HIV and influenza (EC50 values are 15, 32, 30-60, >10 and >80 μM respectively). Orally bioavailable.
Specifications

Specifications

Inhibitor
C12H15N5O3
BMS 200475, SQ 34676
RNA/DNA Polymerase Inhibitors
142217-69-4
277.28
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.