Learn More
Sartorius Entris™ II Advanced Precision Balances
GREENER_CHOICE
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA323I-1S
2237.25 GBP valid until 2025-12-31
Use promo code "24377" to get your promotional price.
 View more versions of this product
Product Code. 16635663
  • £2355.00 / Each
Explore more special offers
Description
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • Accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results by using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Certificate, CSA for Canada
With Internal Calibration
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤1.0 seconds
100/240 V
50/60 Hz
Internal
D-sub connector, 9-pin; USB UTL; USB Device
320 g
1 mg
Electronic Precision Balance
Balance
0.2 A
Class I
Product Suggestions
Product Suggestions
Videos
Videos
  • Sartorius Entris II Advanced Line application peakhold
  • Sartorius Entris II Advanced Line application statistic
  • Sartorius Entris II Advanced Line application mixing
  • Sartorius Entris II Advanced Line application weighing
  • Sartorius Entris II Advanced Line application conversion
  • Sartorius Entris II Advanced Line application percentage weighing
  • Sartorius Entris II Advanced Line application components
  • Sartorius Entris II Advanced Line application counting
  • Sartorius Entris II Advanced Line application pipette smart test
  • Sartorius Entris II Advanced Line application animal weighing
SDS
Documents
Documents
Brochures
Product Guides
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Entris™ II Advanced Precision Balances > 320 g; 1 mg; With Internal Calibration