Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Brand: Eppendorf™ 5804791002
190.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Adapter | |
13 x 5.5/12 mL Tubes | |
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges | |
5.5/12 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
Eppendorf™ S-4-72 Rotor Adapters > For 13 x 5.5/12mL tubes
Spot an opportunity for improvement?Share a Content Correction