missing translation for 'onlineSavingsMsg'
Learn More
ERB 041, Tocris Bioscience™ Product Code.: 18055412

ERB 041, Tocris Bioscience™

Product Code. 18055412
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18055412

Brand: Tocris Bioscience 4276/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent ERβ agonist

Potent ERβ agonist; displays >200-fold selectivity for ERβ over ERα (IC50 values are 5 and 1216 nM for human ERβ and ERα; 3.1 and 620 nM for rat ERβ and ERα; and 3.7 and 750 nM for mouse ERβ and ERα respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524684-52-4
Molecular Formula C15H10FNO3
Molecular Weight (g/mol) 271.247
InChI Key FCXYSEXZEGPLGG-DHDCSXOGSA-N
Synonym prinaberel, unii-a9c8mnf7ca, a9c8mnf7ca, 2-3-fluoro-4-hydroxyphenyl-7-vinyl-1,3-benzoxazol-5-ol, prinaberel usan:inn, 4-2z-7-ethenyl-5-hydroxy-3h-1,3-benzoxazol-2-ylidene-2-fluorocyclohexa-2,5-dien-1-one, prinaberel usan/inn, bidd:er0139, erb hplc, 7-ethenyl-2-3-fluoro-4-hydroxyphenyl benzoxazol-5-olShow MoreShow Less
PubChem CID 5326893
IUPAC Name (4Z)-4-(7-ethenyl-5-hydroxy-3H-1,3-benzoxazol-2-ylidene)-2-fluorocyclohexa-2,5-dien-1-one
SMILES C=CC1=CC(=CC2=C1OC(=C3C=CC(=O)C(=C3)F)N2)O

Specifications

Quantity 10 mg
Formula Weight 271.24
Percent Purity >99%
Chemical Name or Material ERB 041
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ERB 041, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.