Learn More
Notrax ESD Wrist Band Product Code.: 17771918
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
Notrax ESD Wrist Band
Product Code. 17771918
Wrist Band; For Use With: Connect a Worker's Skin to the Ground
Click to view available options
Wrist Band; For Use With: Connect a Worker's Skin to the Ground
Product Code. 17771918
Brand: Notrax 056SWB0100
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
Add to basket
Wrist Bands connect a worker’s skin to ground and are therefore the primary means of controlling static charge on personnel. When properly worn and connected to ground, static charges are safely dissipated from the person.
  • Wrist Bands connect a worker’s skin to ground and are therefore the primary means of controlling static charge on personnel
  • When properly worn and connected to ground, static charges are safely dissipated from the person
  • Meets the requirements of EOS/ESD S1, MIL-STD-1686, and EIA 625
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
Specifications
Product Type Wrist Band
Material Wire
For Use With (Application) Connect a Worker's Skin to the Ground
Product Content Correction
Product Title
Notrax ESD Wrist Band >