Learn More
β-Estradiol 17-acetate, MedChemExpress
MedChemExpress β-Estradiol 17-acetate is a metabolite of estradiol.
Brand:  Medchem Express HY-B0708
Encompass_Preferred
Product Code. 16438426
  • £72.60 / 100mg
Explore more special offers
Description
Description
β-Estradiol 17-acetate is a metabolite of estradiol.
Specifications
Specifications
β-Estradiol 17-acetate
White
C20H26O3
1,3,5(10)-Estratriene-3,17β-diol 17-acetate
C[C@@]1([C@H]2OC(C)=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
314.42
Research
1743-60-8
Research
100 mg
DMSO : 100 mg/mL (318.05 mM; Need ultrasonic)
314.42
95.64%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
β-Estradiol 17-acetate, MedChemExpress