missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™ Product Code.: 15802544
compliance-icons compliance-icons

Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™

Product Code. 15802544
4 L, Glass Bottle
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15802544

Brand: Fisher Chemical A4094

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

A tax-paid 200 proof ethanol that meets USP, EP, and ACS testing specifications to encourage cross-regional collaboration and potentially reduce laboratory inventory for ethanol applications.

  • Meets USP, EP, and ACS testing specifications
  • Consolidate your purchases into one product code and potentially reduce inventory
  • Suitable for use as a cleaner, solvent, or reagent in various applications
  • Packaged in laboratory-friendly sizes
  • Pricing includes tax
  • Combination of compendial test specifications enables use across regions
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Absorbance 0.40 max. at 240nm (USP, EP), 0.30 max at 250-260nm (USP, EP), 0.10 max at 270-340nm (USP, EP)
Boiling Point 78°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max. (ACS)
Water 0.2% max. (ACS)
Formula Weight 46.07
Grade ACS/EP/USP
Chemical Name or Material Ethanol, 200 Proof
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol 200 Proof USP EP ACS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.