missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals Product Code.: 10498450

Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals

Product Code. 10498450
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10498450

Brand: Thermo Scientific Acros 118541000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5405-41-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004545
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
PubChem CID 62572
ChEBI CHEBI:87685
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Colorless
Density 1.0100g/mL
Boiling Point 170°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CO2C2H5
Refractive Index 1.4195 to 1.4215
Quantity 100 g
Beilstein 03, 309
Fieser 14,162
Specific Gravity 1.01
Synonym ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate
Solubility Information Solubility in water: soluble
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 3-hydroxybutyrate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433136 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number118541000,118545000,118540250
Lot NumberA0352907 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog Number118541000,118545000,118540250
Product Identifier
  • Ethyl 3-hydroxybutyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.