missing translation for 'onlineSavingsMsg'
Learn More
Ethyl glycolate, 95%, Thermo Scientific Chemicals Product Code.: 10269740

Ethyl glycolate, 95%, Thermo Scientific Chemicals

Product Code. 10269740
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10269740

Brand: Thermo Scientific Acros 410081000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-50-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00021970
InChI Key ZANNOFHADGWOLI-UHFFFAOYSA-N
Synonym ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl esterShow MoreShow Less
PubChem CID 12184
IUPAC Name ethyl 2-hydroxyacetate
SMILES CCOC(=O)CO

Specifications

Color Colorless
Density 1.0800g/mL
Boiling Point 158.0°C to 159.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula HOCH2COOCH2CH3
Refractive Index 1.4170 to 1.4200
Quantity 100 g
Specific Gravity 1.08
Formula Weight 104.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl glycolate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0354304 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number410080250,410085000,410081000
Lot NumberA0444696 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog Number410080250,410085000,410081000
Lot NumberA0382415 Certificate TypeCertificate of Analysis Date10/03/2022 Catalog Number410080250,410085000,410081000
Lot NumberA0437532 Certificate TypeCertificate of Analysis Date24/12/2021 Catalog Number410080250,410085000,410081000
compliance-icons
Product Identifier
  • Ethyl glycolate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl glycolate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.