missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediaminetetraacetic acid, 99%, Thermo Scientific Chemicals Product Code.: 11489562

Ethylenediaminetetraacetic acid, 99%, Thermo Scientific Chemicals

Product Code. 11489562
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy

Product Code. 11489562

Brand: Thermo Scientific Alfa Aesar A10713.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

Solubility
Soluble in sodium hydroxide, sodium carbonate, ammonia solution, acid and common organic solvents. Slightly soluble in water.

Notes
Stable. Incompatible with copper, copper alloys, nickel, aluminum, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-00-4
Melting Point ∼245°C (decomposition)
Molecular Formula C10H16N2O8
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
MDL Number MFCD00003541
Quantity 250 g
Beilstein 1716295
Merck Index 14,3517
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene
Solubility Information Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble aq. soln..
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol) 292.24
PubChem CID 6049
ChEBI CHEBI:42191
Formula Weight 292.24
Percent Purity 99%
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467038 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog NumberA10713.36,A10713.0I,A10713.30,A10713.0B
Lot NumberA0453447 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog NumberA10713.36,A10713.0I,A10713.30,A10713.0B
Lot NumberW15H027 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog NumberA10713.36,A10713.0I,A10713.30,A10713,A10713.0B
Lot Number10237473 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10713.36,A10713.0I,A10713.30,A10713.0B
Lot Number10239773 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10713.36,A10713.0I,A10713.30,A10713.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.