missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 209°C to 214°C |
Color | Yellow |
Loss on Drying | 1% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97+% |
Packaging | Glass bottle |
Linear Formula | CH3CH2OC(S)SK |
Quantity | 100 g |
Merck Index | 15, 7813 |
Solubility Information | Solubility in water: soluble |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0451206 | Certificate of Analysis | 11/03/2024 | 410505000,410501000,410500250 |
A0437751 | Certificate of Analysis | 05/01/2023 | 410505000,410501000,410500250 |
A0395095 | Certificate of Analysis | 14/12/2022 | 410505000,410501000,410500250 |
A0395460 | Certificate of Analysis | 14/12/2022 | 410505000,410501000,410500250 |
A0402207 | Certificate of Analysis | 13/12/2022 | 410505000,410501000,410500250 |
Safety and Handling

Product Identifier
- Ethylxanthic acid potassium salt
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Ethylxanthic acid potassium salt
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction