missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) chloride hexahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10625082

Europium(III) chloride hexahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10625082
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10625082

Brand: Thermo Scientific Acros 193010050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13759-92-7
Molecular Formula Cl3Eu·6H2O
Molecular Weight (g/mol) 366.4
MDL Number MFCD00149701

Specifications

Melting Point 850.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 5 g
Assay Percent Range 39.0 to 42.5% (Eu) (Complexometry)
Linear Formula EuCl3·6H2O
Fieser 15,186
Merck Index 10, 3854
Solubility Information Solubility in water: soluble
Formula Weight 366.4
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.89
Density 4.8900g/mL
Chemical Name or Material Europium(III) chloride hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465390 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number193010010,193010250,193010050
Lot NumberA0462345 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number193010010,193010250,193010050
Lot NumberA0459754 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number193010010,193010250,193010050
Lot NumberA0461331 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number193010010,193010250,193010050
Lot NumberA0457835 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog Number193010010,193010250,193010050
Product Identifier
  • Europium(III) chloride hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) chloride hexahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.