missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11361039

Europium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11361039
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11361039

Brand: Thermo Scientific Alfa Aesar 011299.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-Show MoreShow Less
PubChem CID 159371
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water.
IUPAC Name dieuropium(3+) trioxidandiide
Formula Weight 351.92
Sensitivity Hygroscopic
Appearance White powder or chunks
Chemical Name or Material Europium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS04E038 Certificate TypeCertificate of Analysis Date13/06/2022 Catalog Number011299.03,011299.14,011299.22,011299.06
Lot NumberQ04I002 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011299.03,011299.14,011299.22,011299.06
Lot NumberX25H041 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011299.03,011299.14,011299.22,011299.06
Lot NumberE24P17 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011299.03,011299.14,011299.22,011299.06
Lot NumberA15W020 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011299.03,011299.14,011299.22,011299.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.