missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals Product Code.: 11478480

Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals

Product Code. 11478480
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11478480

Brand: Thermo Scientific Alfa Aesar 044347.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes, red or blue phosphor in television sets. It shows an useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-Show MoreShow Less
PubChem CID 159371
IUPAC Name dieuropium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Physical Form Powder
Quantity 100 g
Merck Index 14,3904
Solubility Information Insoluble in water.
Formula Weight 351.92
Percent Purity 99.996%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium (III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW19I005 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number044347.14,044347.22,044347.06
Lot NumberY18G033 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number044347.14,044347.22,044347.06
Lot NumberP06A005 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number044347.14,044347.22,044347.06
Lot NumberC01S072 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044347.14,044347.22,044347.06
Lot NumberI11N19 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number044347.14,044347.22,044347.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.