Learn More
Heidolph Instruments Evaporating Flask
For use with rotary evaporators
Brand: Heidolph Instruments 514-72000-00
This item is not returnable.
View return policy
Description
Evaporating flasks come with a standard NS 29/32 joint size but are also available in NS 24/29
Specifications
Borosilicate Glass | |
Evaporating Flask | |
250 mL | |
115/230 VAC |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Heidolph Instruments Evaporating Flask > Capacity: 250 mL
Spot an opportunity for improvement?Share a Content Correction