missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiSf™ CD Medium

Gibco™ ExpiSf™ CD Medium is chemically defined, yeastolate-free, serum-free, protein-free, animal origin-free medium specifically designed for high-density growth, transfection, and baculovirus infection of suspension-adapted Sf9 (Spodoptera frugiperda) insect cells.

Brand:  Gibco™ A3767804

 View more versions of this product

Product Code. 15809126

  • £960.00 / Each
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Delivers high reproducibility and lot-to-lot reliability
  • Facilitates high-level expression of baculovirus-infected cultures, achieving protein yields three times higher than traditional insect expression platforms
  • Supports growth of suspension Sf9 cultures to densities of approximately 2 x 107 cells/mL, with >90% cell viability
  • Supports high-density infection of suspension Sf9 cells with recombinant baculovirus
  • Supports growth, transfection, and baculovirus infection of Sf9 cells in culture formats from 24-deep well plates to >3L in shaker flasks
  • Is a ready-to-use formulation, with no supplementation required
  • manufactured using an ISO 13485-certified process according to 21 CFR 820 medical device current good manufacturing practices (cGMP)

ExpiSf™ CD Medium is a core component of the ExpiSf Expression System. For highest level protein expression, the medium is used in combination with a matched protein expression enhancer, ExpiSf Enhancer, which is supplied as part of the ExpiSf Protein Production Kit (Cat. No. A3767806). Sf21 and High Five™ cells adapted to other commercially-available serum-free media may be adapted for growth in ExpiSf CD Medium; although, sequential adaptation is required and high-density growth isn’t guaranteed. ExpiSf™ CD Medium is not recommended for adherent cell cultures. Cells are typically grown in Erlenmeyer-style flasks on a shaker platform in a non-humidified, non-CO2 cell culture incubator.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 L
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Liquid
ExpiSf™
10 L bag, store in the dark at 2–8°C

Shelf Life: 12 months from date of manufacture

Insect Cells
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers