Learn More
Gibco™ ExpiSf™ CD Medium
Gibco™ ExpiSf™ CD Medium is chemically defined, yeastolate-free, serum-free, protein-free, animal origin-free medium specifically designed for high-density growth, transfection, and baculovirus infection of suspension-adapted Sf9 (Spodoptera frugiperda) insect cells.
£63.75 - £1770.00
Specifications
Classification | Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free |
---|---|
Compatible Cells | Insect Cells |
Format | Liquid |
Shipping Condition | Room Temperature |
Product Line | ExpiSf™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15879116
|
Gibco™
A3767801 |
500 mL |
£63.75
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15889116
|
Gibco™
A3767802 |
1000 mL |
£113.00
Each |
In Stock
Log in to see stock. |
|||||
15899116
|
Gibco™
A3767803 |
6 x 1000 mL |
£608.00
Each |
In Stock
Log in to see stock. |
|||||
15809126
|
Gibco™
A3767804 |
10 L |
£960.00
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15819126
|
Gibco™
A3767805 |
20 L |
£1770.00
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
- Delivers high reproducibility and lot-to-lot reliability
- Facilitates high-level expression of baculovirus-infected cultures, achieving protein yields three times higher than traditional insect expression platforms
- Supports growth of suspension Sf9 cultures to densities of approximately 2 x 107 cells/mL, with >90% cell viability
- Supports high-density infection of suspension Sf9 cells with recombinant baculovirus
- Supports growth, transfection, and baculovirus infection of Sf9 cells in culture formats from 24-deep well plates to >3L in shaker flasks
- Is a ready-to-use formulation, with no supplementation required
- manufactured using an ISO 13485-certified process according to 21 CFR 820 medical device current good manufacturing practices (cGMP)
ExpiSf™ CD Medium is a core component of the ExpiSf Expression System. For highest level protein expression, the medium is used in combination with a matched protein expression enhancer, ExpiSf Enhancer, which is supplied as part of the ExpiSf Protein Production Kit (Cat. No. A3767806). Sf21 and High Five™ cells adapted to other commercially-available serum-free media may be adapted for growth in ExpiSf CD Medium; although, sequential adaptation is required and high-density growth isn’t guaranteed. ExpiSf™ CD Medium is not recommended for adherent cell cultures. Cells are typically grown in Erlenmeyer-style flasks on a shaker platform in a non-humidified, non-CO2 cell culture incubator.
Specifications
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free | |
Liquid | |
ExpiSf™ |
Insect Cells | |
Room Temperature |