missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiSf™ Expression System Starter Kits

The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.

Brand:  Gibco™ A38841

1452.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15859126

  • £1650.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.

The ExpiSf Expression System offers

  • A chemically defined, yeastolate-free, animal origin-free medium for consistent cell growth and protein expression
  • Integrated components that work in concert to deliver three times higher protein yields compared to traditional insect expression platforms
  • Robust production of high-titer, high-quality P0 recombinant baculovirus in suspension culture format without the need for further virus amplification
  • A fast, simplified workflow that allows you to obtain protein in as little as 6–10 days; half the time of traditional protocols
  • Scalable expression that can be applied to cultures from 24-deep well plates to >3 L shake flasks
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insect Cell Protein Expression System
Untagged
1 kit
Kit with Erlenmeyer Flasks
Sf9, SFM Adapted
Polyhedrin
Restriction Enzyme/MCS
Baculovirus

• 1000 mL bottle ExpiSf CD Medium, store in the dark at 2°C to 8°C
• 4 x 1 mL ExpiSf Enhancer, 2°C to 8°C
• 1 mL ExpiFectamine Sf Transfection Reagent, 2°C to 8°C
• 2 x 1 vial ExpiSf9 Cells, store in vapor-phase liquid nitrogen
• 500 mL Opti-MEM, 2°C to 8°C
• 10 μg pFastBac1 vector, 2°C to 8°C
• 4 ng pFastBac1-Gus control vector, 2°C to 8°C
• 5 x 0.1 mL MAX Efficiency DH10Bac Competent Cells, -80°C
• 24 x 125 mL Single-Use Nalgene PETG Erlenmeyer Flasks, store at room temperature

Approved for shipment at Room Temperature or on Wet or Dry Ice
ExpiSf™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.