missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ ExpiSf™ Expression System Starter Kits
The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.
£1650.00
Specifications
Type | Insect Cell Protein Expression System |
---|---|
Delivery Type | Baculovirus |
Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
Cell Line | Sf9, SFM Adapted |
Product Line | ExpiSf™ |
Product Code | Brand | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Price | Quantity & Availability | |||||
15859126
|
Gibco™
A38841 |
Kit with Erlenmeyer Flasks |
£1650.00
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
Description
The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.
The ExpiSf Expression System offers
- A chemically defined, yeastolate-free, animal origin-free medium for consistent cell growth and protein expression
- Integrated components that work in concert to deliver three times higher protein yields compared to traditional insect expression platforms
- Robust production of high-titer, high-quality P0 recombinant baculovirus in suspension culture format without the need for further virus amplification
- A fast, simplified workflow that allows you to obtain protein in as little as 6–10 days; half the time of traditional protocols
- Scalable expression that can be applied to cultures from 24-deep well plates to >3 L shake flasks
Specifications
Insect Cell Protein Expression System | |
Approved for shipment at Room Temperature or on Wet or Dry Ice | |
ExpiSf™ |
Baculovirus | |
Sf9, SFM Adapted |
For Research Use Only. Not for use in diagnostic procedures.