missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Factory Calibration Certificate For 3 Temperatures (37/52/70°C), For Un75Pa
Memmert™ Calibration Certificate with a single measuring point is for three temperatures.
Brand: Memmert™ UN75paD00126
This item is not returnable.
View return policy
Description
- Works calibration certificate for 3 temperatures: +37°C, +52°C, +70°C
- Standard works calibration certificate (measuring point chamber centre) at +80°C
Specifications
Measuring point chamber centre | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate | |
37°C, 52°C, 70°C |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Factory Calibration Certificate For 3 Temperatures (37/52/70°C), For Un75Pa >
Spot an opportunity for improvement?Share a Content Correction