Learn More
Thermo Scientific™ X-Gluc
Effectively screen colonies with X-Gluc, a substrate for ß-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene.
£110.00 - £330.00
Specifications
Detection Method | Colorimetric |
---|---|
Product Type | X-Gluc |
Substrate Properties | Chemical Substrate |
Substrate Type | Beta-Glucuronidase (GUS) Substrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10509140
|
Thermo Scientific™
R0851 |
0.1 g |
£110.00
0.10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10519140
|
Thermo Scientific™
R0852 |
0.5 g |
£330.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).
Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).Recommended for:
- Detection of GUS expression in plant cells and tissues (1)
- Detection of infections caused by E.coli
- Detection of bacterial contamination in food and water samples
Note:
Preparation of a 0.1M stock solution in dimethylformamide or dimethylsulfoxide is recommended.
Specifications
Colorimetric | |
X-Gluc | |
Chemical Substrate | |
Beta-Glucuronidase (GUS) Substrate |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.