Learn More
Thermo Scientific™ X-Gluc
Effectively screen colonies with X-Gluc, a substrate for ß-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene.
Brand: Thermo Scientific™ R0852
234.30 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).
Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).Recommended for:
- Detection of GUS expression in plant cells and tissues (1)
- Detection of infections caused by E.coli
- Detection of bacterial contamination in food and water samples
Note:
Preparation of a 0.1M stock solution in dimethylformamide or dimethylsulfoxide is recommended.
Specifications
Colorimetric | |
0.5 g | |
Beta-Glucuronidase (GUS) Substrate |
X-Gluc | |
Chemical Substrate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.