missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ X-Gluc

Effectively screen colonies with X-Gluc, a substrate for ß-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene.

Brand:  Thermo Scientific™ R0851

78.10 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10509140

  • £110.00 / 0.10g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).

Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).

Recommended for:

  • Detection of GUS expression in plant cells and tissues (1)
  • Detection of infections caused by E.coli
  • Detection of bacterial contamination in food and water samples

Note:

Preparation of a 0.1M stock solution in dimethylformamide or dimethylsulfoxide is recommended.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorimetric
0.1 g
Beta-Glucuronidase (GUS) Substrate
X-Gluc
Chemical Substrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2797000 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0851
Lot Number2817939 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0851
Lot Number2833802 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0851
Lot Number2850581 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0851
Lot Number2922729 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberR0851
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ X-Gluc > 0.1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.