Learn More
Thermo Scientific™ Finnpipette™ Dispenser
GREENER_CHOICE
Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.
£390.00 - £811.00
Specifications
Product Type Analog Bottletop Dispenser
Certifications/Compliance Individual calibration certificate with serial number
Channels 1
View More Specs
Products 6
Product Code Brand Precision Description Price Quantity & Availability  
Product Code Brand Precision Description Price Quantity & Availability  
11448409
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4421120
2µL Range: 0.2-1.0mL, ±6.0µL
£572.00
Each
In Stock
to see stock.
Add to basket
12811171
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
4421130
4 µL Range: 0.4-2.0mL, ±12.0µL
£390.00
Each
In Stock
to see stock.
Add to basket
11468409
View Documents Promotion Details
Thermo Scientific™
4421140
10 µL Range: 1-5mL, ±30.0µL
£537.00
Each
In Stock
to see stock.
Add to basket
11478409
View Documents Promotion Details
Thermo Scientific™
4421150
20µL Range: 2-10mL, ±60.0µL
£537.00
Each
In Stock
to see stock.
Add to basket
11488409
View Documents Promotion Details
Thermo Scientific™
4421160
60 µL Range: 5-30mL, ±180µL
£720.00
Each
In Stock
to see stock.
Add to basket
12326532
View Documents Promotion Details
Thermo Scientific™
4421170
120µL Range: 10-60mL, ±360µL
£811.00
Each
In Stock
to see stock.
Add to basket
Description
Description
Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.
Features:
  • Excellent chemical and thermal resistance
  • Fully autoclavable†
  • Exact self-locking system in the volume setting
  • Dispensing head cap prevents dripping
  • Ergonomic design prevents work fatigue
  • Threaded suction hose for bubble-free suction
Note:
Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.
Warranty: 1 year
Specifications
Specifications
Analog Bottletop Dispenser
Individual calibration certificate with serial number
1
Videos
SDS
Documents
Documents
Product Certifications
Note: Not for use on hot plates.
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ Dispenser