Learn More
Thermo Scientific™ Finnpipette™ Dispenser

Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.
Brand: Thermo Scientific™ 4421120
359.27 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
359.28 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.
Features:
- Excellent chemical and thermal resistance
- Fully autoclavable†
- Exact self-locking system in the volume setting
- Dispensing head cap prevents dripping
- Ergonomic design prevents work fatigue
- Threaded suction hose for bubble-free suction
Note:
Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.

Specifications
Analog Bottletop Dispenser | |
Range: 0.2-1.0mL, ±6.0µL | |
0.2 to 1.0 mL |
Individual calibration certificate with serial number | |
1 | |
2µL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Note: Not for use on hot plates.