Learn More
Thermo Scientific™ Finntip™ Flex Filtered Pipette Tips
Minimize the risk of cross-contamination with these flex filtered pipette tips that combine the ergonomics of a standard pipette tip with the safety of a filter tip.
£84.50 - £369.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
For Use With (Equipment) | Thermo Scientific Finnpipette |
Product Line | FinnTip Flex |
Product Code | Brand | Description | Volume (Metric) | Color Code | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Color Code | Fit for Purpose | Price | Quantity & Availability | ||
11833430
|
Thermo Scientific™
94056980 |
Finntip Flex Filter 10µL, sterile | 10 μL | Pink | Sterile & PCR Ready |
£270.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11843430
|
Thermo Scientific™
94056510 |
Finntip Flex Filter 30µL, sterile | 30 μL | Yellow | Sterile & PCR Ready |
£278.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11853430
|
Thermo Scientific™
94056520 |
Finntip Flex Filter 100µL, sterile | 100 μL | Yellow | Sterile & PCR Ready |
£270.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11863430
|
Thermo Scientific™
94056380 |
Finntip Flex Filter 200µL, sterile | 200 μL | Yellow | Sterile & PCR Ready |
£270.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11852703
|
Thermo Scientific™
94056580 |
Finntip Flex Filter 300µL, sterile | 300 μL | Orange | Sterile & PCR Ready |
£270.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11802713
|
Thermo Scientific™
94056710 |
Finntip Flex Filter 1000µL, sterile | 1000 μL | Blue | Sterile & PCR Ready |
£301.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11862703
|
Thermo Scientific™
94056810 |
Finntip Flex Filter 1200µL, sterile | 1200 μL | Turquoise | Sterile & PCR Ready |
£369.00
Pack of 960 |
In Stock
Log in to see stock. |
||
11475558
|
Thermo Scientific™
94056970 |
Finntip Flex Filter 10mL Ext., sterile | 10 mL | Red | Certified Quality |
£84.50
Pack of 50 |
In Stock
Log in to see stock. |
||
Description
Minimize the risk of cross-contamination with Thermo Scientific™ Finntip™ Flex Filter Pipette Tips, which combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Offered sterile, and free from DNA, DNase and RNase. Ideal for PCR and pipetting of radioactive and infectious samples. The filters, made of an inert nonsealing polyethylene matrix, are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip.
Key Features:
- Excellent for single and multichannel pipetting
- Smooth, uniform inner surface of the tips prevents liquid retention
- Filtered Finntip Flex pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
- Inert and non-sealing polyethylene filter
- Tips are supplied in vacuum sealed, sterilized, 10 × 96 rack (10 mL Extended in bag)
Softer Finntip Flex Tip
- The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
- It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Proven quality
- Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination (excluding 10 mL Extended)
Versatile Tip Range
- Available in volumes: 10, 30, 100, 200, 300, 1000, 1200μL and 10mL Extended (bottle tip)
Immediate traceability
- Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert
Note:
Availability may vary by country.
Specifications
Filtered | |
Sterile | |
Thermo Scientific Finnpipette | |
FinnTip Flex |