missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Taq DNA Polymerase

Allows up to 100 cycles of amplification without loss of activity

Brand:  Fisher BioReagents™ FB6000-75

 View more versions of this product

Product Code. 10104974

  • £3655.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Licensed for use in PCR
  • Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
  • Has a 5«→3« exonuclease activity
  • Available with two different reaction buffers
  • Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
  • Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
  • Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween™ 20, 0.5% Nonidet P40™
  • Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial

Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial
Taq DNA Polymerase
Poly Tube
Standard PCR
Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5%
-20°C
Liquid
5 × 2500 Units
5U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ Taq DNA Polymerase > In Buffer B; 5 x 2500U

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.