missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Taq DNA Polymerase
Allows up to 100 cycles of amplification without loss of activity
Brand: Fisher BioReagents™ FB6000-70
This item is not returnable.
View return policy
Description
- Licensed for use in PCR
- Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
- Has a 5«→3« exonuclease activity
- Available with two different reaction buffers
- Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
- Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
- Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween™ 20, 0.5% Nonidet P40™
- Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial
Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above

Specifications
10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial | |
Taq DNA Polymerase | |
Poly Tube | |
Standard PCR | |
Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5% |
-20°C | |
Liquid | |
2500 Units | |
5U/μL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Taq DNA Polymerase > In Buffer B; 2500U
Spot an opportunity for improvement?Share a Content Correction