Learn More
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator
For monitoring temperature of sensitive samples
Brand:  Fisherbrand™ FB4103
 View more versions of this product
Product Code. 12921161
  • £211.00 / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For monitoring temperature sensitive samples volumes
  • Can be used to monitor volumes from 1 - 1000mL
  • Temperature monitoring thermocouple bead on the end of a hermetically sealed wire
Specifications
Specifications
Thermocouple Probe
Ideal for monitoring temperature of samples >1mL and indirect sonication methods
FB 705 Sonic Dismembrator™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator > Flexible Thermocouple Probe