Learn More
Fisherbrand™ Traceable™ Sound Level Meter
Meter receives sound through a 1/2 in. condenser microphone.
Brand:  Fisherbrand™ 93676-46
Product Code. 11794266
  • £419.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Meter receives sound through a 1/2 in. condenser microphone.
Resolution is 0.1dB with accuracy of ± 2 db. Response time can be set for fast (0.2 seconds) or slow (1.5 seconds). MAX HOLD function retains highest reading recorded for later display. Recorder jack permits capturing readings (AC and DC mV/dB) for use with most chart recorders.
  • Three decibel ranges for greater precision
  • Adjustable response times
  • Measures low (30.0 to 80.0dB), medium (50.0 to 100.0dB), high (80.0 to 130.0dB)
Order Info
Dimensions: 3L x 1W x 3 in.H (25.4 x 7.6 x 2.5cm). Net weight: 91/2 oz. (269g). Also supplied are 9V alkaline battery and carrying case.
Certifications
Supplied with individually serial-numbered ISO 7025 certificate indicating traceability to NIST.
Compliance
Meets IEC 651 Type 2 standards by providing weightings for both A (frequencies from 500 to 10kHz) and C (frequencies from 30 to 10kHz).
Specifications
Specifications
Traceable certificate, IEC 651 Type 2, OSHA
Sound Level Meter
3 in.
9 V alkaline battery, carrying case, 0.5 in. condenser microphone, recorder jack, threaded tripod insert
7.62 cm
0.1 dB
9.5 oz.
1 in.
3 x 1 x 3 in. (7.62 x 2.54 x 7.62 cm)
500 Hz to 10 KHz or 30 Hz to 10 KHz
7.62 cm
3 in.
35 to 80 dB low, 50 to 100 dB medium, 80 to 130 dB high
0.2 sec. fast, or 1.5 sec. slow
269.32 g
2.54 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Traceable™ Sound Level Meter > Sound Level Meter