missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this N-hydroxysuccinimide modification reagent.

Brand:  Thermo Scientific™ PG82072

 View more versions of this product

Product Code. 15225763

  • £2460.00 / 10g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Amine-Carboxyl
217.13
Upon receipt store at 4°C.
Ambient
Chemical Labeling
Carbodiimide
Powder
Water
Yes
No
No
No
Pierce™
Heterobifunctional
Short (<10 Å)
10 g
Standard, Single-use, Premium-grade
Crosslinker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA405366 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberPG82072
Lot NumberYK385815 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberPG82072
Lot NumberYF339208 Certificate TypeCertificate of Analysis Date07/07/2023 Catalog NumberPG82072
Lot NumberSPECS Certificate TypeCertificate of Analysis Date22/06/2023 Catalog NumberPG82072
Lot NumberWI309750 Certificate TypeCertificate of Analysis Date11/10/2021 Catalog NumberPG82072
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide) > 10g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.