Learn More
Thermo Scientific™ Pierce™ Premium Grade EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride)
Optimize applications where product integrity and risk minimization are paramount with this high-quality formulation of carbodiimide crosslinker.
Brand: Thermo Scientific™ PG82079
39.83 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
54.86 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo ScientificTM PierceTM Premium Grade EDC, our highest quality formulation of this popular carbodiimide crosslinker.
- The highest quality formulation of this popular carbodiimide crosslinker, specially characterized for applications where product integrity and risk minimization are paramount
- Compared to the standard grade product, Premium Grade EDC provides more clearly defined quality and product support
- A water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation
- In the presence of N-hydroxysulfosuccinimide (sulfo-NHS), EDC can be used to convert carboxyl groups to amine-reactive sulfo-NHS esters
- Store dessicated at -20°C
Features of Premium Grade EDC
- High quality – identity and purity confirmed by several tests
- Product integrity – enhanced level of testing and characterization compared to standard grade
- Lot retention – ample supply of past lots retained to ensure future process testing
- Change management – Change Control Notification (CCN) service
- Consistent manufacture – batch-specific manufacturing documentation review
Applications
- Conjugate carboxyl and amino groups among peptides and proteins
- Couple haptens to immunogenic carrier proteins
- Immobilize peptide antigens to affinity-purify antibodies
- Create NHS-activated, amine-reactive labeling compounds
- Crosslink proteins to carboxyl coated beads or surfaces
- Activate nanoparticles with amine-reactive sulfo-NHS esters

Specifications
Amine-Carboxyl | |
191.7 | |
0.0 Å | |
No | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
1 g | |
Standard, Single-use, Premium-grade | |
Crosslinker |
No | |
No | |
Upon receipt store desiccated at -20°C. | |
Room Temperature | |
Chemical Labeling | |
Carbodiimide | |
Solid | |
Water | |
Yes |
Safety and Handling
- Pierce Premium Grade, EDC
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : N'-(ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.