Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Racks
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80862997
 View more versions of this product
Product Code. 15736949
  • £81.15 / Pack of 2
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Mixes 96-well plates or deepwell blocks
  • Requires Insert Retainer (02-215-391)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.5 and 0.5 mL
Microtube Foam rack
Foam
Digital and analog mini vortex mixer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Vortex Mixer Accessories: Foam Racks > Foam rack; For microtubes