Learn More
Fisherbrand™ Amber Soda Glass Wide Mouth Packer Bottles with White Polypropylene/Polyvinyl Lined Cap
Ideal for storage of dry powders.
£12.17 - £724.53
Specifications
Color Amber
Includes Certificate of analysis
Material Soda Lime Glass
Closure Color White
Closure Included Yes
View More Specs
Products 11
Product Code Brand Capacity Closure Size Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity Closure Size Diameter (Metric) Height (Metric) Price Quantity & Availability  
11724259
View Documents
Fisherbrand™
02911852A
0.2 oz., 6 mL 20-400 mm 24 mm 44 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11734259
View Documents
Fisherbrand™
02911799A
0.5 oz., 15 mL 28-400 mm 31 mm 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11754319
View Documents
Fisherbrand™
02911432A
1 oz., 30mL 28-400 mm 37 mm 65 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11758749
View Documents
Fisherbrand™
02911433A
2 oz., 60 mL 33-400 mm 44 mm 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11744259
View Documents
Fisherbrand™
02911448A
2 oz., 60 mL 33-400 mm 66 mm 119 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11754259
View Documents
Fisherbrand™
02911449A
4 oz., 125 mL 38-400 mm 54 mm 95 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11774319
View Documents
Fisherbrand™
02911435A
8 oz., 250 mL 45-400 mm 66 mm 119 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11784319
View Documents
Fisherbrand™
02911436
16 oz., 500 mL 53-400 mm 80 mm 146 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11734319
View Documents
Fisherbrand™
02911451A
16 oz., 500 mL 53-400 mm 80 mm 146 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11744319
View Documents
Fisherbrand™
02911452A
32 oz., 1000 mL 53-400 mm 99 mm 178 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11778849
View Documents
Fisherbrand™
02911437A
32 oz., 1000 mL 53-400 mm 99 mm 178 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Wide mouth for bottles allows easy accessibility for removal of contents
  • Custom cleaned containers and caps are free of visible particulates done using deionised water
  • Certified clean containers are cleaned as per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
Specifications
Specifications
Amber
Soda Lime Glass
Yes
Wide Mouth Packer Bottle
Screw Cap
Certificate of analysis
White
Round
Polypropylene