Learn More
Fisherbrand™ Sterile Cell Strainers
Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
£143.00 - £144.86
Specifications
Mesh Material | Nylon |
---|---|
Disposable | Single-use |
Material | Polypropylene |
Product Type | Cell Strainer |
Sterility | Sterile |
Product Code | Brand | Color | Mesh Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Mesh Size | Price | Quantity & Availability | |||||
11597522
|
Fisherbrand™
22363548 |
White | 70 μm |
£143.00
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
11517532
|
Fisherbrand™
22363549 |
Yellow | 100 μm |
£144.86
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
11587522
|
Fisherbrand™
22363547 |
Blue | 40 μm |
£143.00
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
Description
Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Rapidly isolate primary cells- Fast, simple alternative to gauze filtration
- Reliably dissociates cells from clumps or primary tissues
- Consistently obtain a uniform single-cell suspension
- Ideal for preparation of:
- Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
- Specimens for primary cell cultures and immunogens
- Freezing stocks
- Filtering agglutinative proteins produced in inactivation serum
- Strong nylon mesh with evenly spaced pores for consistent results
- Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
- Includes an extended lip for easier handling with forceps
- Color-coded polypropylene frame with tab enables easy handling and identification
- Individually packed, ready to use and disposable
Specifications
Nylon | |
Polypropylene | |
Sterile |
Single-use | |
Cell Strainer | |
Conical Tubes, 50 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.