Learn More
Fisherbrand™ Round Bottom Disposable Borosilicate Glass Tubes with Plain End
Use these tubes for tissue culture and clinical chemistry applications. Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
£76.00 - £368.00
Product Code | Brand | Diameter (Metric) | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | |||
12347279
|
Fisherbrand™
|
10 mm | 75 mm | 4 mL |
£76.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
11517403
|
Fisherbrand™
|
12 mm | 75 mm | 6 mL |
£82.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
11527403
|
Fisherbrand™
|
13 mm | 100 mm | 10 mL |
£100.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
11537403
|
Fisherbrand™
|
15 mm | 85 mm | 13 mL |
£115.58
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
11547403
|
Fisherbrand™
|
16 mm | 100 mm | 15 mL |
£118.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
11557403
|
Fisherbrand™
|
16 mm | 125 mm | 19 mL |
£151.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
11577403
|
Fisherbrand™
|
16 mm | 150 mm | 23 mL |
£171.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
11587403
|
Fisherbrand™
|
18 mm | 150 mm | 28 mL |
£203.30
Pack of 500 |
In Stock
Log in to see stock. |
|||
11597403
|
Fisherbrand™
|
20 mm | 150 mm | 36 mL |
£180.00
Pack of 500 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||
11507413
|
Fisherbrand™
|
25 mm | 150 mm | 55 mL |
£368.00
Pack of 500 |
In Stock
Log in to see stock. |
|||
Description
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass. These disposable borosilicate glass tubes are perfect for tissue culture and clinical chemistry applications.
- 10 and 12mm O.D. sizes are available
- Packaged in shrink-wrapped trays of 125 or 250 to prevent contamination or breakage
Specifications
Borosilicate Glass | |
Yes |