missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.

Brand:  Fisherbrand™ 90008-87

98.08 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11749745

  • £133.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.

  • Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
  • Glass bottle filled with glycol mimics vaccine temperature and fits in trays
  • Probe is sealed in glycol solution which insulates sensor when refrigerator door is opened
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
  • Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • High-impact, chemical-resistant ABS plastic case
  • Temperature range: -50 to +70°C/-58 to +158°F
  • Resolution: 1° accuracy: ±1C
  • Battery:AA Alkaline (1)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It provides traceability to standards provided by NIST.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    29

    Hours

    19

    Minutes

    20

    Seconds

    17
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer > Refrigerator/Freezer Thermometer/Alarm w/5mL glycol bottle attachment

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.