missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Wide-Form Porcelain Crucibles
Alert:
Heating and cooling rates should not exceed 200°C per hour and maximum use temp of 1100°C
Description
Withstand temperatures up to 1150°C (2102°F)
- Glazed inside and out except for outside bottom surface and rim
Order Info
Covers available separately
Specifications
Porcelain | |
1150°C | |
17g | |
41mm | |
Crucible |
Glazed | |
2mm | |
17 mL | |
26mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Wide-Form Porcelain Crucibles > 41 x 26 x 2mm; 17g; Case wt.: 1.7kg; Capac.: 17mL
Spot an opportunity for improvement?Share a Content Correction