Learn More
Saint-Gobain Versilon™ R-3400 UV Resistant Acid Transfer Tubing
Ideal for virtually any permanent or temporary chemical transfer application, Saint Gobain Performance Plastics™ Versilon™ R-3400 UV Resistant Acid Transfer Tubing combines suppleness and flexibility with resistance to a wide range of chemicals.
£20.55 - £92.92
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | |||
10616385
|
Saint-Gobain
AAE00002 |
3.2 mm | 1.6 mm | 0.8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12386819
|
Saint-Gobain
AAE00017 |
9.6 mm | 6.4 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10461915
|
Saint-Gobain
AAE00012 |
8 mm | 4.8 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Features and Benefits
- Black in color to protect light-sensitive fluids
- Compatible with a wide range of chemicals
- Ozone resistant
- High temperature rating for excellent burn resistance
- Low compression set to minimize permanent deformation
Typical Applications
- Acid and caustic transfer
- Corrosive diffusable gases
- Fertilizer and pesticide dispensing
- Electrical insulation lines
- Ink and adhesive dispensing
Specifications
Each | |
Black | |
Flexible |