Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
£171.21 - £203.42
Specifications
Style Upright
Color Silver
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Type Price Quantity & Availability  
11786469
View Documents
Thermo Scientific™
224134
13.0 cu. ft. Freezers Shelf Kit
£171.21
Each
Estimated Shipment: 25-09-2025
to see stock.
Add to basket
11756479
View Documents
Thermo Scientific™
224135
17.3 cu. ft. Freezers Shelf Kit
£179.69
Each
Estimated Shipment: 27-08-2025
to see stock.
Add to basket
11766479
View Documents Promotion Details
Thermo Scientific™
224158
23.0 cu. ft. Freezers Shelf Kit
£203.42
Each
In Stock
to see stock.
Add to basket
11786649
View Documents
Thermo Scientific™
224180
28 cu. ft. Freezers Shelf Kit
£172.91
Each
Estimated Shipment: 25-09-2025
to see stock.
Add to basket
Description
Description
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
  • Stainless steel
  • Kits include mounting hardware
Specifications
Specifications
Upright
Silver
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Forma™ Freezer Replacement Shelves