missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Formaldehyde, 4% in PBS Product Code.: 15413179

Thermo Scientific Chemicals Formaldehyde, 4% in PBS

Product Code. 15413179
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15413179

Brand: Thermo Scientific Chemicals J60401.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses.

Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehydeShow MoreShow Less
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Packaging Plastic bottle
Quantity 250 mL
Solubility Information Soluble in water,ethanol,acetone,and ether.
Chemical Name or Material Formaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP15L506 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberJ60401.AP,J60401.AK
Lot NumberP16L506 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberJ60401.AP,J60401.AK
Lot NumberM11L548 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberJ60401.AP,J60401.AK
Lot NumberW23K503 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ60401.AP,J60401.AK
Lot NumberX01K535 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ60401.AP,J60401.AK
compliance-icons compliance-icons
Product Identifier
  • Formaldehyde, 4% in PBS
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : formaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Formaldehyde, 4% in PBS >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.