missing translation for 'onlineSavingsMsg'
Learn More
Paraformaldehyde Solution, 4% in PBS, Thermo Scientific Chemicals Product Code.: 15670799

Paraformaldehyde Solution, 4% in PBS, Thermo Scientific Chemicals

Product Code. 15670799 Shop All Thermo Scientific Products
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 15670799

Brand: Thermo Scientific J19943.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Physical Form Solution
Packaging Plastic bottle
Quantity 1 L
Concentration 4% in PBS
MDL Number MFCD00133991
Chemical Name or Material Paraformaldehyde Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number245173 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberJ19943.K2
Lot Number233480 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberJ19943.K2
Lot Number235885 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ19943.K2
Lot Number235575 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ19943.K2
Lot Number195273 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog NumberJ19943.K2
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde solution, 4% in PBS, Affymetrix/USB
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Paraformaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Paraformaldehyde Solution, 4% in PBS, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.