missing translation for 'onlineSavingsMsg'
Learn More
Formamide, 99+%, for spectroscopy, Thermo Scientific Chemicals Product Code.: 10627201

Formamide, 99+%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10627201
500 mL, Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10627201

Brand: Thermo Scientific Acros 410765000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88Show MoreShow Less
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.0°C to 3.0°C
Absorbance 0.01 at 360nm, 0.01 at 400nm, 0.05 at 300nm, 0.2 at 280nm, 0.5 at 275nm
Density 1.1330g/mL
Boiling Point 210.0°C
Quantity 500 mL
Flash Point 175°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4460 to 1.4480
Assay Percent Range 99+%
Specific Gravity 1.133
Linear Formula HCONH2
Viscosity 3.75 mPa.s (20°C)
Beilstein 02, 26
Water 0.03% max. (Coulometric)
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Formula Weight 45.04
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material Formamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464380 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number410760025,410760010,410765000
Lot NumberA0453479 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number410760025,410760010,410765000
Lot NumberA0429131 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number410760025,410760010,410765000
Lot NumberA0442055 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number410760025,410760010,410765000
Lot NumberA0394260 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number410760025,410760010,410765000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formamide, 99+%, for spectroscopy, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.